Pre-Cleaned Straight-Sided Jars, Polyethylene, Environmental Express
Catalog # 23050-048
Supplier: Environmental Express
Small Business Enterprise
Specifications
- Cap size:89-400
- Capacity:1 L (33.8 oz.)
- Description:Level III Jar
- Cat. no.:23050-048
- Supplier No.:BPC3240
Specifications
About this item
Natural high-density poylethylene jars include a polypropylene cap and F-217 PE liner.
All containers meet one of the three levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.
Customers Who Bought This Also Bought