VWR® Talon® Support Stand, Cast Iron, Tripod Base
Catalog # 12985-076
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Ø:15.2 cm (6")
- Cat. no.:12985-076
Specifications
About this item
Support stand base is constructed of cast iron with black enamel finish.
- Cast iron construction with black enamel finish
- Tripod base
- Built-in support rod holder with locking knob
- For use with VWR® Talon® lab frames
Base features a built-in support rod holder with locking knob that accepts a 1.3 cm (1/2") diameter support rod (sold separately).
Customers Who Bought This Also Bought