ContactUsLinkComponent
Pre-Cleaned Glass Jars with Open-Top Cap and Septa, Clear, Environmental Express
Catalogue # 23050-202
Fournisseur:  Avantor
Small Business Enterprise
Pre-Cleaned Glass Jars with Open-Top Cap and Septa, Clear, Environmental Express
Catalogue # 23050-202
Fournisseur:  Avantor
Small Business Enterprise
Spécifications
  • Cap Size:
    48-400
  • Capacity:
    125 ml (4.2 oz.)
  • Description:
    Level I Quality Assured Jars
  • Cat. no.:
    23050-202
  • Supplier No.:
    APC1994
Spécifications
À propos de cet article
These clear glass jars come with an open-top polypropylene cap and PTFE liner.
All containers meet one of three levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance. Level I pre-cleaning is the same as Level II, but instead of a Statement of Compliance, a Certificate of Analysis is provided indicating that the containers received full EPA quality assurance treatment.