Pre-Cleaned Wide-Mouth Glass Jars, Clear, Environmental Express®
Fournisseur: Environmental Express
Certificats
A propos de cet article
All containers meet one of two levels of pre-cleaning.
Amber glass jars include polypropylene closure and PTFE liner.
Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody sealed, lot number labeled for traceability, and a Certificate of Analysis for EPA quality assurance treatment.
Customers Who Bought This Also Bought