Sample Vial Convenience Kits, Borosilicate Glass with Open-Top Screw Caps and Septa, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Unassembled convenience kits include clear, borosilicate glass vials with polypropylene, open-top screw caps, and natural PTFE/clear silicone septa.
Ordering information: Cartons contain shrink-wrapped vials and separately packaged caps and septa in plastic bags.
Customers Who Bought This Also Bought