MPC Series Plastic Couplings, Colder Products Company
Fournisseur: Colder Products
Certificats
A propos de cet article
Couplings allow rapid disconnect and reconnect with one hand and without the need for tools
Couplings are available in polysulfone (amber tint) or polycarbonate (purple tint) construction. Coupling inserts include seals constructed of clear silicone. All products meet USP Class VI criteria.
They feature an ergonomic thumb latch and an audible click when connected, assuring the user of a secure connection. Couplings can be rotated when connected, reducing the inconvenience of tube kinks. These items are sterilizable and reusable, yet economical enough to allow disposability.
Certifications: All products meet USP Class VI criteria and feature BSE/TSE-free materials (ADCF).
Spécifications
- Catalog No:
- 16001-590
- 16001-598
- 16001-596
- 16001-594
- 16001-592
- 16001-600
- 16001-588
- 16001-586
- Max. operating pressure:
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Vacuum to 60 psi (4.2 bar)
- Operating temperature:
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)
- Polycarbonate –40 to 121°C (–40 to 250°F), Polysulfone –40 to 148.9°C (–40 to 300°F)