ContactUsLinkComponent
Fuchs-Rosenthal Counting Chamber, Hausser Scientific
Fuchs-Rosenthal Counting Chamber, Hausser Scientific
# de catalogue 15170-230
Fournisseur:  Hausser
Fuchs-Rosenthal Counting Chamber
Fuchs-Rosenthal Counting Chamber, Hausser Scientific
# de catalogue 15170-230
Fournisseur:  Hausser
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    15170-230
  • L×W×D:
    75 x 32 x 4.5 mm (215/16x11/4x3/16")
  • Supplier No.:
    3720
Spécifications
A propos de cet article
Hemacytometer for counting cells in cerebrospinal fluid, etc
Cell depth: 0.2 mm. Fuchs-Rosenthal ruling with sixteen 1mm squares each subdivided into 16 smaller squares. One piece construction. Supplied with two cover glasses. Without case.
Customers Who Bought This Also Bought