Septa, Silicone, Wheaton
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Red silicone septa for use with aluminum seals.
Item 224168 is a specially molded septa with a PTFE inset. The sealing surface of butyl and PTFE effect a more positive seal than plain PTFE faced septa.
Customers Who Bought This Also Bought