Aluminum Seals, Tear-Off, WHEATON®
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Seals can be punctured by syringe or completely removed and contents poured from bottle
Shelf packs of 100.
Customers Who Bought This Also Bought