Phase Hemacytometer, Hausser Scientific
Phase Hemacytometer
Catalog # 15170-263
Supplier: Hausser
Fournisseur #: 3200
Jump to:
Product Details & Documents
Phase hemacytometer is used with a phase microscope for accurately counting red and white blood cells, platelets, dust/yeast clusters and specimens in spinal and other body fluids
Triple dividing lines show clearly which cells lie within the counting area. A “V” cut into the plateau makes filling easy and reduces the possibility of overflow into the moat. Each plateau features enhanced Neubauer rulings 0.1mm below the cover glass.
Ordering information: Supplied with two reusable cover glasses.
Spécifications
- Désignation:Phase Hemacytometer