A propos de cet article
VWR® Maximum Protection Non-Skid Shoe Covers are the cleanest and most durable within the entire VWR Protection line of apparel
These non-skid shoe covers are the cleanest and most durable within the entire VWR Protection line of apparel. Engineered to optimize skid-resistance, barrier protection, cleanliness, and comfort. Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications. Serged seams. Available in white or blue.
- Non-skid, abrasion resistant sole provides maximum traction on wet and smooth surfaces
- Superior coefficient of friction
- Impervious material construction
- Clean, low-linting fabric
- Serged seams
Shoe covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Engineered to optimize skid-resistance, barrier protection, cleanliness, and comfort. Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Abrasion resistance: 1000 (sole)
Spécifications
- Catalog No:
- 414004-489
- 414004-488
- 414004-483
- 414004-482
- 414004-857
- 414004-487
- 414004-486
- 414004-480
- 414004-481
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Bacterial filtration efficiency:
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- >99.9%
- Particle shedding (Helmke Drum):
- Level I
- Level I
- Level I
- Level I
- Level I
- Level I
- Level I
- Level I
- Level I
- Water resistance:
- Pass
- Pass
- Pass
- Pass
- Pass
- Pass
- Pass
- Pass
- Pass
- Tensile strength:
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- 20.3 (Filling)
- Tearing strength:
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- 13.7 (Widthwise Yarns)
- Weight:
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²
- 58.58 g/m²