Tubes for Micro Rack Tube System, Greiner Bio-One
Fournisseur: Greiner Bio-One
Certificats
A propos de cet article
Tubes are designed to work with the Micro Rack tube system, which allows safe and secure multichannel dispensing into these disposable tubes.
- Disposable tube design
- Translucent
The translucent 1.3 ml polypropylene tubes are available in sterile or non-sterile formats.
Customers Who Bought This Also Bought