PicoSolutions Horizontal Wind Energy Standard
Fournisseur: PICOSOLUTIONS GROUP INC SE
New Product
Certificats
A propos de cet article
Explore horizontal wind energy with a DIY hands-on turbine model.
- 6th grade to college level tailored activities for different learning stages
- Subjects spanning sustainable energy, math, aerodynamics, geometry
- Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind features a DC motor for beginners and an AC generator with 12+ experiments for advanced learners. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine, with blade adapters for custom blades made from dowels or 3D printing.
Ordering information: Educational support bundle: Upgrade available for enhanced learning with support from a certified virtual teacher mentor and DESS course. Future-ready skills: The DESS course enriches the kit by integrating design, entrepreneurship, and STEM, fostering creativity, innovation, and problem-solving abilities.
Spécifications
- Catalog No:
- 470355-360
- 470355-300
- 470355-376
- 470355-350
- 470355-318
- 470355-340
- Dimensions:
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"