Accessories for Safety Funnels with Lid, 'MARCO' V2.0
Fournisseur: SCAT AMERICAS, INC.
New Product
Certificats
A propos de cet article
Replacement parts for SCAT 'MARCO' V2.0 safety funnels, which can be closed safely and prevent the danger of ignition.
- Suitable for all models of the MARCO series
- Available in conductive and non-conductive versions
Stainless steel sieve to catch small objects such as stirrers from HPLC equipment.
Customers Who Bought This Also Bought