Shooting Start Jar, Stone Meteorites
# de catalogue 470355-900
Fournisseur: AEROLITE METEORITES INC SE
New Product
Spécifications
- Description:Shooting star jar
- Size:100 - 150 g wt.
Spécifications
A propos de cet article
Actuals hooting starts from outer space.
- NWA XXX
- Stone meteorite
- 100% guaranteed to be authentic and as described
- Collected in the sahara desert
- Contained in a glass vial with a cork lid
A shooting star is one of the most spectacular, yet fleeting, occurrences in the night sky. Legend holds that if you close your eyes and make a wish on a shooting star, your wish will come true. Now you can hold the rare cosmic power of shooting stars in the palm of your hand with our exclusive jar of shooting stars. Feel the wonder of the cosmos as you ponder the unearthly beauty of these incredible rocks from outer space. They originated in the outer crust of a planet or asteroid.
Packaging: Vial dimensions are 56×36×36 mm.
Customers Who Bought This Also Bought