Meteorite Dust Galaxy Collection
# de catalogue 470355-902
Fournisseur: AEROLITE METEORITES INC SE
New Product
Spécifications
- Description:Meteorite dust galaxy collection
- Size:<1 g wt.
Spécifications
A propos de cet article
Five glass vials containing autheticated iron, stone, lunar, martian, and vestal meteorite dust.
- 100% guaranteed to be authentic and as described
- Economical galactic starter bundle
These quaint glass vials contain genuine meteorite 'dust'. The dust is collected in our preparation laboratory, when a larger specimen is cut into slices–similar to how sawdust is created. We capture everything from the saw, because meteorites are precious.
Because no two meteorites or impactities are the same, yours will be similar but not exact.
Packaging: Vial dimensions are 21×16 mm.
Customers Who Bought This Also Bought