Visitors from Space: Mixed Meteorite, Pack of 4
# de catalogue 470355-912
Fournisseur: AEROLITE METEORITES INC SE
New Product
Spécifications
- Description:Meteorite, pack of 4
Spécifications
A propos de cet article
Economical starter pack of meteorites and impactities.
- All three major meteorite types included
- 100% guaranteed to be authentic and as described
- Convient packaging to share amongst a small group of students, display, and store
- Colorful insert included
This fantastic starter set includes stone, iron, and stony-iron meteorites and an impactite. Impact glasses are created by ancient meteorite events on earth. Because no two meteorites or impactities are the same, yours will be similar but not exact.
Packaging: Package is 109×109×20 mm.
Customers Who Bought This Also Bought