KIMBLE® KONTES® Allihn Condensers, DWK Life Sciences
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificats
A propos de cet article
This borosilicate glass Allihn condenser are used in many refluxing operations.
- Standard taper drip joint at bottom and a standard taper outer joint at top
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought