µltraAmp™ PCR Cap Strips, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
Polypropylene cap strips are ideal for real-time PCR applications
They prevent evaporation during thermal cycling or storage and are optimized to fit 96-well plates. Each strip contains eight caps. Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Frequently Bought Together