Flame-Resistant Sleeves, Kevlar® with Cotton Lining, Wells Lamont®
Fournisseur: Wells Lamont
Certificats
A propos de cet article
These cut and flame-resistant 2-ply sleeves are constructed of Kevlar with cotton lining for optimal protection and comfort.
- Yellow
- Four sizes available
- Kevlar construction for protection
- Cotton lining for comfort
- Flame resistant
The yellow sleeves can be placed in the dryer, not exceeding 150°F (67°C), after washing.
The sleeves are available in four sizes.
Certifications: ANSI Cut Level 3, A3.
Caution: Caution Information: Cut-resistant sleeves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.
Spécifications
- Catalog No:
- 80091-790
- 80091-802
- 80091-774
- 80091-780
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Kevlar® with cotton lining
- Kevlar® with cotton lining
- Kevlar® with cotton lining
- Kevlar® with cotton lining