KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These KIM caps are designed to fit culture tubes with outer diameters ranging from 13 mm to 38 mm.
- Autoclavable
- Color coded
Customers Who Bought This Also Bought