VWR® Dual-Position Caps, PE
Fournisseur: VWR International
Certificats
A propos de cet article
White caps for use with VWR® and most popular brand disposable culture tubes.
- Polyethylene fabrication
- Dual-position caps
- Compatible with most disposable culture tubes
Caps permit aspiration of cultures.
Spécifications
- Catalog No:
- 60819-091
- 60818-463
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact