Premium Back Support Belt, OK-1® Safety, OccuNomix
Fournisseur: Occunomix International, LLC
Certificats
A propos de cet article
Featuring strong, power-knit nylon and elastic construction, these belts are designed for people who lift average weight loads or are involved in repetitive motion at work
- Strong, power-knit nylon and elastic construction
- For average weight loads or repetitive motion
- Lumbar pad molds comfortably
- Sturdy double-closure system overlaps twice for superior abdominal support
The lumbar pad molds comfortably into the lower lumbar region of the spine, while the sturdy double closure system overlaps twice for superior abdominal support.
Ordering information: All come with 2-way stretch, 4cm (11/2") wide suspenders, featuring breakaway swivel suspender clips that assure the suspenders detach only when you want them to.
Customers Who Bought This Also Bought