NALGENE® Polypropylene Screw Caps, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Linerless caps are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable.
Ordering information: Bulk packed.
Customers Who Bought This Also Bought