Nichrome Resistance Wire, Arcor Electronics
Fournisseur: Arcor Electronics
Certificats
A propos de cet article
Used for making heating wires and inoculating loops
60% nickel, 16% chromium, 24% iron. Maximum operating temperature: 1150°C (2100°F). Bare, 1/4 lb. spool.
Customers Who Bought This Also Bought