Sealing Disc, PTFE, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Convert ordinary caps into non-contaminating closures at a fraction of the cost with a Polyethylene F217 Foam Disc.
- Easy-to-clean
- Easy to remove
- Easy to replace
Just press the disc into your existing closure.
Customers Who Bought This Also Bought