White Polypropylene Screw Caps, PE Foam Liner, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Polypropylene caps with PE Foam liners are known for good impact strength, and pliability.
- Cost effective
- Excellent sealing characteristics
These polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, and household oils.
Customers Who Bought This Also Bought