VWR® Standard Standard Sealed and Septum Caps
Fournisseur: VWR International
Certificats
A propos de cet article
Caps are manufactured from medical grade, FDA approved, high-density polypropylene.
- High-density polypropylene construction
- Reusable and recyclable
- Autoclavable
- Supplied nonsterile
They are autoclavable, reusable, and recyclable
Septum caps are available with a gray butyl rubber insert. They are ideal for injectables or containment of pathogens.
VWR® polycarbonate media bottles, vials, and culture tubes are supplied with a standard seal cap. Blue is the standard color for all caps, although custom colors are available upon request.
All caps are linerless with an integrally-molded seal ring to ensure leakproof performance. They are manufactured in a Class 10K cleanroom environment and are supplied nonsterile.
Customers Who Bought This Also Bought