White Metal Screw Caps, Plastisol Liner, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
White metal caps are impact-resistant and exhibit the widest range of temperature tolerances
Caps feature Plastisol liners which offer excellent resistance to mild acids. For use with glass or plastic wide mouth bottles. Not autoclavable.
Customers Who Bought This Also Bought