Black Phenolic Screw Caps, Polyethylene Liner, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Caps with solid PE liners offer good chemical resistance and a low moisture vapor transmission rate and are for great polish remover and higher concentration acids & alkalis
These black phenolic screw caps are made of phenolic resin.
Good for non-oil products filled at room temperature. Because this lining system is paperless, your samples will not be contaminated with fiber or paper particles from the closure.
Customers Who Bought This Also Bought