WHEATON® Black Plastic Caps for Media Bottles, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Solid-top lined caps are suitable for a variety of applications
Choose from rubber-, fluoropolymer resin-, polyethylene-, or metal foil-lined caps. Rubber-lined caps are for culture work. Fluoropolymer resin-lined caps are for maximum product safety. Both rubber- and fluoropolymer resin-lined caps are autoclavable. Polyethylene-lined caps are designed for distilled water, analytical standards, and reagents. Metal foil-lined caps are for organic solvents.
Customers Who Bought This Also Bought