PMC Series Plastic Couplings, Colder Products Company
Fournisseur: Avantor
Certificats
A propos de cet article
These quick disconnect couplings cover a wide range of general purpose applications from chemical analyzers to medical devices
They feature integral terminations, which results in fewer leak points, shorter assemblies, and faster installations. An audible click assures the user of a secure connection. Couplings include a thumb latch for one-hand connection and disconnection without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues. Components and valves are constructed of acetal thermoplastic, with stainless steel latches, springs, and pins. O-rings are made of Buna-N. Couplings are produced in a natural white tint.
Ordering information: Polypropylene couplings for greater chemical resistance are also available; contact your Avantor sales representative.
Spécifications
- Catalog No:
- 16001-854
- 16001-960
- 16001-886
- 16001-988
- 16001-828
- 16001-912
- 16001-954
- Max. operating pressure:
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Operating temperature:
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous