PLC Series Plastic Couplings, Colder Products Company
Supplier: Colder Products
Certificates
About this item
These quick disconnect couplings are injection molded from acetal thermoplastic, making them highly durable and resistant to most chemical solutions
Both the external and valve springs are constructed of stainless steel. Couplings feature integral terminations which results in fewer leak points, shorter assemblies, and faster installations. An audible click assures the user of a secure and reliable connection. Couplings have a thumb latch for rapid disconnect and reconnect with one hand, without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues.
Ordering information: Polypropylene couplings for greater chemical resistance are available; contact your Avantor sales representative.
Specifications
- Catalog No:
- 16001-650
- 16001-806
- 16001-648
- 16001-706
- 16001-644
- 16001-796
- Max. operating pressure:
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Vacuum to 120 psi (8.3 bar)
- Operating temperature:
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous
- –40 to 82°C (–40 to 180°F) continuous