Drying Rack, Vinyl-Coated Steel Wire, Accurate Wirecraft Company
# de catalogue 60996-009
Fournisseur: Accurate Wirecraft
Spécifications
- Length:181/2"
- Width:71/4"
- Height:19"
- Cat. no.:60996-009
- Weight:4.5 lbs.
- Supplier No.:901807BK
Spécifications
A propos de cet article
For draining and drying test tubes, beakers, flasks, and other laboratory glassware
Can be used flat, hung on wall, or placed upright on table or sink. Saves glassware from chipping and breaking. The vinyl coating is resistant to many chemicals and oils. Holds 90 pieces of glassware. Contains seventy-two 7.6cm (3") pins and eighteen 12.7cm (5") pins. Made of strong, extruded, steel wire, spot-welded throughout. All drainage is caught in the rustproof pan.
Customers Who Bought This Also Bought