EP Scientific Septa, PTFE/Silicone, Thermo Scientific
# de catalogue 89039-972
Fournisseur: Thermo Scientific
Spécifications
- Ø:24 - 400 mm
- Description:Pre-cleaned standard flat disc PTFE/silicone septa
- Thickness:1.5 mm (0.060")
- Cat. no.:89039-972
- Supplier No.:S24-400-S2
Spécifications
A propos de cet article
Standard flat disc PTFE/silicone septa for use with EP Scientific EPA Vials.
- Fits the standard 24-414 open top cap
Certifications: Processed to meet EPA Performance Based Specifications for Volatile Organic Analysis.
Ordering information: Other sizes are available. Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought