All-Aluminum Inner Seals, West Pharmaceutical Services
Fournisseur: West Pharmaceutical
Certificats
A propos de cet article
For use with injection and infusion bottles, dental cartridges, X-ray imaging solutions, oral liquids, and opthalmic, veterinary, and agricultural applications
Plain aluminum ferrule seals feature lacquered shell and center-holes for crimping stoppers to glass vials. Seals have a turned-in-edge and are capable of high-speed processing.
Customers Who Bought This Also Bought