Draining/Drying Rack, Wood, Bennett Wood
# de catalogue 60998-004
Fournisseur: Bennett Wood Specialties
Spécifications
- Length:181/2"
- Width:20"
- Height:20"
- Cat. no.:60998-004
- Supplier No.:6790
Spécifications
A propos de cet article
Used for draining and drying test tubes, beakers, cylinders.
Ordering information: Complete with 18 5" pins and 72 3" pins.
Customers Who Bought This Also Bought