Synthware Distilling Trap Adapters, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Distilling trap adapter is placed above flask to prevent carryover of reactions that have a tendency to foam.
- Top [ST] joint
- Bottom [ST] joint
Adapter features an upper [ST] joint size of 24/40 and capacity of 100 ml.
Customers Who Bought This Also Bought