Adapter, Conversion, Ace-Thred, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Adapters are designed for use with a connecting adapter to connect vials or flasks to rotary evaporators.
- Ace-Thred bottom
- Top [ST] outer joint
- Manufactured from borosilicate glass
Adapters feature either a #15 or #25 Ace-Thred bottom and a variety of [ST] outer top joint sizes.
Customers Who Bought This Also Bought