DuPont™ Tyvek® IsoClean® Coveralls with Standard Collar
Fournisseur: DuPont
Certificats
A propos de cet article
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Standard laydown collar
- Set sleeve design
- Zipper closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Collar and closure allow for easy donning and doffing.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Packaging: Clean Processed garments are individually packaged.
Spécifications
- Catalog No:
- CA80076-744
- CA80076-750
- CA80076-786
- 10016-064
- 10016-066
- 10016-068
- 10016-070
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
Spécifications
Informations connexes