VWR® CryoPro® Rack Systems, BR Series
Fournisseur: VWR International
Certificats
A propos de cet article
Combining the convenience of box vial storage with the economy of a low nitrogen loss design, the BR Series provides the perfect solution for medium to large storage needs.
- Vapor temperatures of -185 ºC are achieved with as little as 2" of liquid
- Vial storage in boxes (sold separately) allows for quick and accurate inventorying and recovery of specimens
- The BR-1 model can hold 25-cell cardboard or plastic boxes
- The BR-2, BR-3, and BR-4 models can hold 100-cell cardboard or plastic boxes
Optional sturdy five-wheel rollerbases are available for safely moving full tanks.
CryoRetriever claw-style tool is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks. Exclusive sealed center shaft prevents hazardous spurting of LN2 that can occur with a hollow shaft retrieval claw.
VWR® CryoPro® Rack Systems are supplied with a two-year parts and labor warranty (plus an additional three-year vacuum warranty).
Ordering information: An optional low liquid level alarm helps protect against accidental warmups. The phase separator can be attached to the stainless steel transfer hose for increased efficiency and safety when transferring liquid nitrogen. There are also polycarbonate or fibreboard boxes, cryogenic gloves, 6" liquid nitrogen transfer hose and a liquid level measuring stick are available as optional accessories.
Delivery: The BR-1 and BR-4 models include 7 racks. The BR-2 and BR-3 models include 5 racks.
Spécifications
- Catalog No:
- 55709-208
- 55709-210
- 55709-212
- 55709-214
- Model:
- BR-1
- BR-2
- BR-3
- BR-4
- Evaporation rate:
- 0.39 L/Day
- 0.85 L/Day
- 0.99 L/Day
- 0.99 L/Day
- Liquid nitrogen capacity:
- 47 L
- 61 L
- 121 L
- 175 L
- Neck int.Ø:
- 12.7 cm (5")
- 21.6 cm (8¹/₂")
- 21.6 cm (8¹/₂")
- 21.6 cm (8¹/₂")
- Vial capacity:
- 875×2 ml vials
- 2500×2 ml vials
- 5000×2 ml vials
- 7000×2 ml vials
- Weight Empty:
- 16.2 kg (34 lb.)
- 34.2 kg (76 lb.)
- 51 kg (114 lb.)
- 56 kg (125 lb.)
- Weight Full:
- 54 kg (120 ib.)
- 75 kg (166 ib.)
- 140 kg (311 ib.)
- 189 kg (419 ib.)