Wheaton M-T Vial File® Storage Cases, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Clear polystyrene cases provide compact, easily accessible means of vial storage.
For Vial File cases that include glass vials, see 66011-653 series.
- Foam inserts provide extra protection for the vials
- Hinged lids with alphanumeric indexing keep samples organized
Clear polystyrene cases provide compact, easily accessible means of vial storage.
Reusable cases store up to sixty 2 mL or forty 4 mL vials in individual partitions.
For Vial File cases that include glass vials, see 66011-653 series.
Delivery: Each storage case is supplied with one index card and one partition.
Packaging: Cases are packed in corrugated outer cartons suitable for storage.
Customers Who Bought This Also Bought