Nalgene® Cryoware Marker Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Permanent markers with extra-fine tips for marking vials, cardboard, and plastic boxes
Excellent for permanent, colorful identification of cryogenic samples. Ink will not fade at ultra-low temperatures. Markers are not alcohol resistant.
Frequently Bought Together