VWR® Counter-Pen™ Colony Counter
# de catalogue 21800-060
Fournisseur: VWR International
Spécifications
- Description:VWR® Counter-Pen Colony Counter
- Dimensions:15.2L x 1.7dia. cm (6 x 2/3")
- Weight:28.4 g (1 oz.)
- Cat. no.:21800-060
Spécifications
A propos de cet article
Made of durable ABS plastic, this combination marker and digital counter is designed for the lab and plant.
- LCD display
- Counts up to 99,999
- Simultaneous touch, sound, and count
Use the unit to count cell growth on Petri dishes, inventory parts, and tally samples received. Each time a black mark is made with the felt-tipped pen, the unit automatically sounds a beep, and the LCD displays the count. Touch, sound, and count occur simultaneously. Even when interrupted, marks hold your place and the counter holds your total, making the unit virtually error-proof. Unit features audio/visual counts to 99,999 with reset button, on/off switch, and pen cap.
Customers Who Bought This Also Bought