KONTES® U-Shaped Spargers, Purge and Trap, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Designed for use with Tekmar® 2000 and 3000 purge and trap concentrators and with the 2016 and 2032 autosamplers
Fritted spargers are recommended for potable water and wastewater samples; fritless spargers are recommended for water, soils, polymers, pharmaceuticals, and foods. Mount: 1.3 cm (1/2").
Customers Who Bought This Also Bought