VWR® Signature™ Boot Covers made with DuPont™ Tyvek® Material
Fournisseur: VWR International
Certificats
A propos de cet article
Boot covers provide a high level of protection and comfort for all cleanroom applications
Boot covers feature a 45.7cm (18") high upper with elastic at the top of the leg openings. They are supplied with ankle ties.
- High level of protection and comfort for all cleanroom applications
- Nonwoven material protects against dry particulates and nonhazardous liquids
- Soles made from PVC, coarsely textured interior prevents slipping inside and outside the garment
- 45.7cm (18") high upper with elastic at leg openings
- Supplied with ankle ties
Boot covers provide a high level of protection and comfort for all cleanroom applications
Boot covers feature a 45.7cm (18") high upper with elastic at the top of the leg openings. They are supplied with ankle ties.
The nonwoven material offers protection against dry particulates as well as nonhazardous liquids. All boot covers are equipped with Gripper™ soles for slip-resistant performance. These soles are made from PVC and are combined with a coarsely textured interior to prevent slipping inside the garment as well as outside.
Spécifications
- Catalog No:
- CA89013-154
- CA89013-156
- 89013-158
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom