Grip Handle Weights, Class F, Troemner
Fournisseur: Troemner
Certificats
A propos de cet article
Used primarily to test commercial weighing devices
Calibrated according to NIST specification for Class F, as defined by NIST Handbook 105-1, which specifies weights used by government officials to verify commercial weighing devices. Also acceptable for use verifying scales with <=0.1% accuracy.
Certifications: Available with NIST traceable certificate, which provides documentation for basic quality control requirements. NIST/NVLAP certificates are available for more stringent requirements.
Customers Who Bought This Also Bought