Weighing Pans, Aluminum, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Disposable Aluminum Weigh Dishes and Pans can be used as evaporating dishes, dust covers, or balance pans.
Avoid contamination problems; these dishes have no oil residue or vinyl coating. These crimped wall dishes feature increased rigidity and large tab. Great for routine weighing and moisture testing, as well as an embedding mold. Disposable weigh dishes are ideal for dispensing, sampling, and mixing.
Customers Who Bought This Also Bought