Dura Shoe Covers
# de catalogue 21923-220
Fournisseur: Interworld
Spécifications
- Garment application:Laboratory
- Shoe size:Universal
- Color:Lilac
- Cat. no.:21923-220
- Supplier No.:10135-109-02
Spécifications
A propos de cet article
Covers made of spunbound polypropylene bound to an exterior layer of compressed polyethylene (CPE) film
Seamless design effectively contains particulates from street shoes. Heavily textured and snug fitting for better slip resistance. Dimensions: 40.6Lx16.5Hcm (16x7"). Relaxed opening: 12.7cm (5"). Stretched opening: 27cm (105/8").
Customers Who Bought This Also Bought