About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
- Laydown collar
- Open wrists
- Extends below hips
- Front snap closure
- Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Frocks feature body-length jacket design with front snap closures for easy donning and doffing. Ideal for applications that do not require the full body coverage of a coverall. Frocks also feature a laydown collar.
Specifications
- Catalog No:
- CA10010-080
- CA10010-068
- CA10010-072
- 10016-918
- CA10010-070
- 10016-920
- CA10010-074
- 10016-922
- CA10010-076
- CA10010-078
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory